cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Leverandør: Apollo Scientific

OR938498-10G OR938498-1G OR938498-5G
APOSOR938498-10GEA 12325 NOK
APOSOR938498-10G APOSOR938498-1G APOSOR938498-5G
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₄O₃
Lagringstemperatur: Romtemperatur
CAS-nummer: 73094-35-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR