2,2-Dimethylvaleronitrile 95%

Leverandør: Apollo Scientific

Synonymer: 2,2-Dimethylpentanenitrile

OR939158-5G
APOSOR939158-5GEA 8400 NOK
APOSOR939158-5G
2,2-Dimethylvaleronitrile 95%
2,2-Dimethylvaleronitrile
Formel: C₇H₁₃N
MW: 111.19 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 20654-47-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR