2,2-Dimethylcyclopentanone 95%

Leverandør: Apollo Scientific

Synonymer: 2,2-Dimethylcyclopentan-1-one

OR939516-25G
APOSOR939516-25GEA 2675 NOK
APOSOR939516-25G
2,2-Dimethylcyclopentanone 95%
2,2-Dimethylcyclopentanone
Formel: C₇H₁₂O
MW: 112.172 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00049164
CAS-nummer: 4541-32-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR