Methyl quinoline-5-carboxylate 98%

Leverandør: Apollo Scientific

OR940019-1G
APOSOR940019-1GEA 1550 NOK
APOSOR940019-1G
Methyl quinoline-5-carboxylate 98%
Methyl quinoline-5-carboxylate
Formel: C₁₁H₉NO₂
MW: 187.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06204270
CAS-nummer: 16675-62-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR