3-(Diethylamino)-2,2-dimethylpropan-1-ol 95%

Leverandør: Apollo Scientific

OR940572-5G
APOSOR940572-5GEA 1025 NOK
APOSOR940572-5G
3-(Diethylamino)-2,2-dimethylpropan-1-ol 95%
3-(Diethylamino)-2,2-dimethylpropan-1-ol
Formel: C₉H₂₁NO
MW: 159.269 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085710
CAS-nummer: 39067-45-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR