4-Bromo-2-(piperidin-1-ylsulphonyl)anisole 97%

Leverandør: Apollo Scientific

Synonymer: 1-((5-Bromo-2-methoxyphenyl)sulphonyl)piperidine

OR9412-1G
APOSOR9412-1GEA 3125 NOK
APOSOR9412-1G
4-Bromo-2-(piperidin-1-ylsulphonyl)anisole 97%
4-Bromo-2-(piperidin-1-ylsulphonyl)anisole
Formel: C₁₂H₁₆BrNO₃S
MW: 334.23 g/mol
Smeltepunkt: 95…99 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02043682
CAS-nummer: 295360-83-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR