(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)methanol 95%

Leverandør: Apollo Scientific

Synonymer: Bicyclo[4.2.0]octa-1,3,5-trien-3-ylmethanol

OR941854-1G
APOSOR941854-1GEA 2575 NOK
APOSOR941854-1G
(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)methanol 95%
(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)methanol
Formel: C₉H₁₀O
MW: 134,175 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09842342
CAS-nummer: 53076-11-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR