5-Bromo-2-(bromomethyl)benzonitrile 95+%

Leverandør: Apollo Scientific

OR941937-250MG
APOSOR941937-250MGEA 2075 NOK
APOSOR941937-250MG
5-Bromo-2-(bromomethyl)benzonitrile 95+%
5-Bromo-2-(bromomethyl)benzonitrile
Formel: C₈H₅Br₂N
MW: 274,94 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09261033
CAS-nummer: 156001-53-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR