(S)-1-(2,4-Dichlorophenyl)ethanamine 95%

Leverandør: Apollo Scientific

OR942322-1G
APOSOR942322-1GEA 2600 NOK
APOSOR942322-1G
(S)-1-(2,4-Dichlorophenyl)ethanamine 95%
(S)-1-(2,4-Dichlorophenyl)ethanamine
Formel: C₈H₉Cl₂N
MW: 190,07 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06761787
CAS-nummer: 133492-69-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR