(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%

Leverandør: Apollo Scientific
Warning

Synonymer: (S)-2,2-Dimethylcyclopropanecarboxamide

OR9436-25G
APOSOR9436-25GEA 735 NOK
APOSOR9436-25G
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Formel: C₆H₁₁NO
MW: 113.1600 g/mol
Smeltepunkt: 135…137 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00216614
CAS-nummer: 75885-58-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR