Ethyl-3-cyanopropanoate 95%

Leverandør: Apollo Scientific

Synonymer: Ethyl 3-cyanopropanoate

OR946046-1G
APOSOR946046-1GEA 1500 NOK
APOSOR946046-1G
Ethyl-3-cyanopropanoate 95%
Ethyl-3-cyanopropanoate
Formel: C₆H₉NO₂
MW: 127,14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01695650
CAS-nummer: 10137-67-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR