N-Methyl-N-(4-nitrophenyl)acetamide 95%

Leverandør: Apollo Scientific

OR947248-1G
APOSOR947248-1GEA 5550 NOK
APOSOR947248-1G
N-Methyl-N-(4-nitrophenyl)acetamide 95%
N-Methyl-N-(4-nitrophenyl)acetamide
Formel: C₉H₁₀N₂O₃
MW: 194.19 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00035947
CAS-nummer: 121-95-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR