Ac-Leu-nhme 95%

Leverandør: Apollo Scientific

OR947967-1G
APOSOR947967-1GEA 7975 NOK
APOSOR947967-1G
Ac-Leu-nhme 95%
Ac-Leu-nhme
Formel: C₉H₁₈N₂O₂
MW: 186.255 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00038290
CAS-nummer: 32483-15-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR