4-Nitro-D-phenylalanine

Leverandør: Apollo Scientific
Warning

Synonymer: H-D-Phe(4-NO2)-OH, H-D-Phe(4-NO₂)-OH

OR9505-100G OR9505-25G OR9505-5G
APOSOR9505-100GEA 1200 NOK
APOSOR9505-100G APOSOR9505-25G APOSOR9505-5G
4-Nitro-D-phenylalanine
4-Nitro-D-phenylalanine
Formel: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066016
CAS-nummer: 56613-61-7
EINECS: 213-446-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR