2-Hydroxy-6-(2-methylphenyl)pyridine 96%

Leverandør: Apollo Scientific

OR954652-1G
APOSOR954652-1GEA 4475 NOK
APOSOR954652-1G
2-Hydroxy-6-(2-methylphenyl)pyridine 96%
2-Hydroxy-6-(2-methylphenyl)pyridine
Formel: C₁₂H₁₁NO
MW: 185,222 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11876169
CAS-nummer: 300395-32-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR