3-Hydroxycinnamic acid methyl ester 95%

Leverandør: Apollo Scientific

OR955453-1G
APOSOR955453-1GEA 5375 NOK
APOSOR955453-1G
3-Hydroxycinnamic acid methyl ester 95%
3-Hydroxycinnamic acid methyl ester
Formel: C₁₀H₁₀O₃
MW: 178,185 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09259949
CAS-nummer: 66417-46-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR