Methyl quinoline-8-carboxylate 95%

Leverandør: Apollo Scientific

OR955684-1G OR955684-5G
APOSOR955684-1GEA 3400 NOK
APOSOR955684-1G APOSOR955684-5G
Methyl quinoline-8-carboxylate 95%
Methyl quinoline-8-carboxylate
Formel: C₁₁H₉NO₂
MW: 187.195 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06203774
CAS-nummer: 40245-26-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR