7-Methyl-1,3-benzoxazol-2-amine 95%

Leverandør: Apollo Scientific

OR956063-1G
APOSOR956063-1GEA 9925 NOK
APOSOR956063-1G
7-Methyl-1,3-benzoxazol-2-amine 95%
7-Methyl-1,3-benzoxazol-2-amine
Formel: C₈H₈N₂O
MW: 148,162 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD19374461
CAS-nummer: 1268154-07-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR