2-Carboxy-5-chlorophenylboronic acid 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Borono-4-chlorobenzoic acid

OR9568-1G
APOSOR9568-1GEA 3950 NOK
APOSOR9568-1G
2-Carboxy-5-chlorophenylboronic acid 97%
2-Carboxy-5-chlorophenylboronic acid
Formel: C₇H₆BClO₄
MW: 200,384 g/mol
Smeltepunkt: 236…238 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08235091
CAS-nummer: 874290-67-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR