2-Chloroindole 95%

Leverandør: Apollo Scientific

Synonymer: 2-Chloro-1H-indole

OR957032-1G
APOSOR957032-1GEA 6625 NOK
APOSOR957032-1G
2-Chloroindole 95%
2-Chloroindole
Formel: C₈H₆ClN
MW: 151,593 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04117990
CAS-nummer: 7135-31-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR