3-Chloro-N-methylbenzamide 95%

Leverandør: Apollo Scientific

OR957050-1G
APOSOR957050-1GEA 9925 NOK
APOSOR957050-1G
3-Chloro-N-methylbenzamide 95%
3-Chloro-N-methylbenzamide
Formel: C₈H₈ClNO
MW: 169,608 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01214034
CAS-nummer: 18370-10-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR