2-(3-Hydroxyphenoxy)acetonitrile 95%

Leverandør: Apollo Scientific

OR957488-1G
APOSOR957488-1GEA 4475 NOK
APOSOR957488-1G
2-(3-Hydroxyphenoxy)acetonitrile 95%
2-(3-Hydroxyphenoxy)acetonitrile
Formel: C₈H₇NO₂
MW: 149.147 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11181964
CAS-nummer: 107223-69-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR