tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate 98%

Leverandør: Apollo Scientific

OR957715-1G
APOSOR957715-1GEA 2425 NOK
APOSOR957715-1G
tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate 98%
tert-Butyl 4-hydroxy-1H-pyrazole-1-carboxylate
Formel: C₈H₁₂N₂O₃
MW: 184,192 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23110529
CAS-nummer: 1394947-77-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR