2-Mercapto-5-methoxybenzimidazole 98%

Supplier: Apollo Scientific
Warning

OR9581-100G OR9581-25G OR9581-500G
APOSOR9581-100GEA 305 NOK
APOSOR9581-100G APOSOR9581-25G APOSOR9581-500G
2-Mercapto-5-methoxybenzimidazole 98%
2-Mercapto-5-methoxybenzimidazole
Formula: C₈H₈N₂OS
Melting Pt: 255…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00134581
CAS Number: 37052-78-1
EINECS: 253-326-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR