Methyl quinoline-2-carboxylate 97%

Leverandør: Apollo Scientific

OR959530-5G
APOSOR959530-5GEA 1075 NOK
APOSOR959530-5G
Methyl quinoline-2-carboxylate 97%
Methyl quinoline-2-carboxylate
Formel: C₁₁H₉NO₂
MW: 187.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00160630
CAS-nummer: 19575-07-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR