{4-[2-(Dimethylamino)ethoxy]phenyl}methanol

Leverandør: Apollo Scientific
Danger

OR9618-1G
APOSOR9618-1GEA 4475 NOK
APOSOR9618-1G
{4-[2-(Dimethylamino)ethoxy]phenyl}methanol
{4-[2-(Dimethylamino)ethoxy]phenyl}methanol
Formel: C₁₁H₁₇NO₂
MW: 195,258 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08435911
CAS-nummer: 131028-54-1
FN: 2922
ADR: 8+6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR