4-(Hydroxymethyl)benzonitrile 98%

Leverandør: Apollo Scientific

Synonymer: 4-Cyanobenzyl alcohol

OR962541-25G OR962541-100G
APOSOR962541-25GEA 1250 NOK
APOSOR962541-25G APOSOR962541-100G
4-(Hydroxymethyl)benzonitrile 98%
4-(Hydroxymethyl)benzonitrile
Formel: C₈H₇NO
MW: 133.15 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00870633
CAS-nummer: 874-89-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR