EDTA dinatrium salt 98%

Leverandør: Apollo Scientific

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

OR964961-1KG
APOSOR964961-1KGEA 4475 NOK
APOSOR964961-1KG
EDTA dinatrium salt 98%
EDTA dinatrium salt
Formel: C₁₀H₁₄N₂Na₂O₈
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR