2-Mercapto-5-methyl-1,3,4-thiadiazole

Leverandør: Apollo Scientific
Warning

OR9661-1KG OR9661-250G
APOSOR9661-1KGEA 2150 NOK
APOSOR9661-1KG APOSOR9661-250G
2-Mercapto-5-methyl-1,3,4-thiadiazole
2-Mercapto-5-methyl-1,3,4-thiadiazole
Formel: C₃H₄N₂S₂
Smeltepunkt: 182…189 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00038349
CAS-nummer: 29490-19-5
EINECS: 249-667-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR