Methyl-4-amino-3-bromobenzoate 98%

Leverandør: Apollo Scientific
Danger

Synonymer: Methyl 4-amino-3-bromobenzoate

OR9763-1G
APOSOR9763-1GEA 180 NOK
APOSOR9763-1G
Methyl-4-amino-3-bromobenzoate 98%
Methyl-4-amino-3-bromobenzoate
Formel: C₈H₈BrNO₂
MW: 230,06 g/mol
Smeltepunkt: 105…109 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01861385
CAS-nummer: 106896-49-5
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR