3-Butoxybenzoic acid

Leverandør: Apollo Scientific

OR9793-1G OR9793-500MG OR9793-5G
APOSOR9793-1GEA 1725 NOK
APOSOR9793-1G APOSOR9793-500MG APOSOR9793-5G
3-Butoxybenzoic acid
3-Butoxybenzoic acid
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01143605
CAS-nummer: 93351-38-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR