4-Chloro-3-nitropyridine 95%

Leverandør: Apollo Scientific
Danger

OR9995-25G
APOSOR9995-25GEA 750 NOK
APOSOR9995-25G
4-Chloro-3-nitropyridine 95%
4-Chloro-3-nitropyridine
Formel: C₅H₃ClN₂O₂
MW: 158,54 g/mol
Smeltepunkt: 33…40 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD02683025
CAS-nummer: 13091-23-1
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR