3,5-Dichloro-α,α,α-trifluorotoluene 97%

Leverandør: Apollo Scientific
Danger

PC0797-25G
APOSPC0797-25GEA 1700 NOK
APOSPC0797-25G
3,5-Dichloro-α,α,α-trifluorotoluene 97%
3,5-Dichloro-α,α,α-trifluorotoluene
Formel: C₇H₃Cl₂F₃
MW: 215 g/mol
Kokepunkt: 173…174 °C
Smeltepunkt: –13…–12 °C
Tetthet: 1.47 g/cm³ (25 °C)
Flash punkt: 65 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00203929
CAS-nummer: 54773-20-5
FN: 1760
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR