1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol

Leverandør: Apollo Scientific

Synonymer: 1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)-1,4-diazepan-1-yl]propan-2-ol

PC10115-500MG
APOSPC10115-500MGEA 8475 NOK
APOSPC10115-500MG
1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol
1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol
Formel: C₁₁H₁₈F₆N₂O₂
MW: 324,263 g/mol
Smeltepunkt: 77…79 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02082052
CAS-nummer: 477847-09-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR