4-Nitro-2-(trifluoromethyl)benzyl alcohol

Leverandør: Apollo Scientific
Warning

Synonymer: (4-Nitro-2-(trifluoromethyl)phenyl)methanol, 2-(Hydroxymethyl)-5-nitrobenzotrifluoride, [4-Nitro-2-(trifluoromethyl)phenyl]methanol

PC10461-1G PC10461-250MG PC10461-5G
APOSPC10461-1GEA 1850 NOK
APOSPC10461-1G APOSPC10461-250MG APOSPC10461-5G
4-Nitro-2-(trifluoromethyl)benzyl alcohol
4-Nitro-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆F₃NO₃
MW: 221.133 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11847130
CAS-nummer: 859027-06-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR