Ammoniumheksafluorosilikat 98%

Leverandør: Apollo Scientific
Danger

Synonymer: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

PC1103-1KG
APOSPC1103-1KGEA 2250 NOK
APOSPC1103-1KG
Ammoniumheksafluorosilikat 98%
Ammoniumheksafluorosilikat
Formel: H₈F₆N₂Si
MW: 178.15 g/mol
Tetthet: 2.011 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010887
CAS-nummer: 16919-19-0
EINECS: 240-968-3
FN: 2854
ADR: 6.1 III
Merck Indeks: 13,00529

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR