Antimony(V) fluoride compound with graphite (50:50%)

Leverandør: Apollo Scientific
Danger

PC1131-25G
APOSPC1131-25GEA 0 NOK
APOSPC1131-25G
Antimony(V) fluoride compound with graphite (50:50%)
Antimony(V) fluoride compound with graphite

Easier to handle form of SbF5. Doesn't fume in air. For the exchange of organic chlorides with fluorine.J. Chem. Soc., Chem. Commun., 815 (1973).

Formel: C:SbF₅
MW: 228.75 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00236643
CAS-nummer: 59839-60-0
FN: 2923
ADR: 8+6.1 II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR