Antimony(III) fluoride 98%

Leverandør: Apollo Scientific
Danger

PC1140-1KG PC1140-250G PC1140-50G
APOSPC1140-1KGEA 4300 NOK
APOSPC1140-1KG APOSPC1140-250G APOSPC1140-50G
Antimony(III) fluoride 98%
Antimony(III) fluoride

A mild fluorinating agent; Fluorination activity may be increased by addition of Br2, Cl2, or SbCl 5. Swarts, Bull. Acad. Roy. Belg., 1892, 24, 309

Formel: F₃Sb
Kokepunkt: 319 °C
Smeltepunkt: 292 °C
Tetthet: 4,38 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011218
CAS-nummer: 7783-56-4
EINECS: 232-009-2
FN: 2923
ADR: 8+6.1,II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR