Bariumfluorid 99%

Leverandør: Apollo Scientific
Danger

PC1170-25G
APOSPC1170-25GEA 450 NOK
APOSPC1170-25G
Bariumfluorid 99%
Barium fluoride
Formel: BaF₂
Kokepunkt: 2260 °C
Smeltepunkt: 1353 °C
Tetthet: 4,89 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003450
CAS-nummer: 7787-32-8
EINECS: 232-108-0
FN: 1564
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR