Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate

Leverandør: Apollo Scientific
Warning

Synonymer: Ethyl [4-(trifluoromethyl)benzoyl]acetate, Ethyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate

PC1854-5G
APOSPC1854-5GEA 1175 NOK
APOSPC1854-5G
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Ethyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Formel: C₁₂H₁₁F₃O₃
MW: 260.209 g/mol
Kokepunkt: 248…249 °C
Tetthet: 1.27 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03424818
CAS-nummer: 106263-53-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR