4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-3-fluorophenylboronic acid pinacol ester

PC200440-25G
APOSPC200440-25GEA 1825 NOK
APOSPC200440-25G
4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%
4-Cyano-3-fluorobenzeneboronic acid pinacol ester
Formel: C₁₃H₁₅BFNO₂
MW: 247.076 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD16996321
CAS-nummer: 870238-67-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR