5-(4-Bromo-2-fluorophenyl)-3-methyl-1,2,4-oxadiazole

Leverandør: Apollo Scientific

PC200494-500MG
APOSPC200494-500MGEA 6900 NOK
APOSPC200494-500MG
5-(4-Bromo-2-fluorophenyl)-3-methyl-1,2,4-oxadiazole
5-(4-Bromo-2-fluorophenyl)-3-methyl-1,2,4-oxadiazole
Formel: C₉H₆BrFN₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09261267
CAS-nummer: 845306-17-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR