4-(4-Fluorophenyl)piperidine hydrochloride 97%

Leverandør: Apollo Scientific

PC201277-5G
APOSPC201277-5GEA 1950 NOK
APOSPC201277-5G
4-(4-Fluorophenyl)piperidine hydrochloride 97%
4-(4-Fluorophenyl)piperidine hydrochloride
Formel: C₁₁H₁₅ClFN
MW: 215.7000 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01632528
CAS-nummer: 6716-98-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR