Cobalt(II) fluoride, vannfri 99%

Leverandør: Apollo Scientific
Danger

Synonymer: Cobalt difluoride, Cobaltous fluoride

PC2080-100G PC2080-25G PC2080-1G
APOSPC2080-100GEA 2075 NOK
APOSPC2080-100G APOSPC2080-25G APOSPC2080-1G
Cobalt(II) fluoride, vannfri 99%
Cobalt(II) fluoride
Formel: CoF₂
MW: 96.93 g/mol
Kokepunkt: 1400 °C
Smeltepunkt: 1200 °C
Tetthet: 4.46 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010941
CAS-nummer: 10026-17-2
EINECS: 233-061-9
FN: 2923
ADR: 8+6.1 III
Merck Indeks: 13,02465

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR