rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

Leverandør: Apollo Scientific

PC210000-1G
APOSPC210000-1GEA 6625 NOK
APOSPC210000-1G
rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid
rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid
Formel: C₁₁H₈FNO₂S
MW: 237.25 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09907872
CAS-nummer: 433283-22-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR