(Aza((4-fluorophenyl)amino)methylene)methane-1,1-dicarbonitrile

Leverandør: Apollo Scientific

Synonymer: 2-[2-(4-Fluorophenyl)hydrazono]malononitrile

PC210059-1G
APOSPC210059-1GEA 1700 NOK
APOSPC210059-1G
(Aza((4-fluorophenyl)amino)methylene)methane-1,1-dicarbonitrile
(Aza((4-fluorophenyl)amino)methylene)methane-1,1-dicarbonitrile
Formel: C₉H₅FN₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00170788
CAS-nummer: 94853-74-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR