4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-N'-hydroxybenzenecarboximidamide

Leverandør: Apollo Scientific

PC210063-1G
APOSPC210063-1GEA 1700 NOK
APOSPC210063-1G
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-N'-hydroxybenzenecarboximidamide
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-N'-hydroxybenzenecarboximidamide
Formel: C₁₃H₉ClF₃N₃O₂
MW: 331,678 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00107676
CAS-nummer: 263161-24-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR