(4-fluorophenyl)(pyridin-4-yl)methanol

Leverandør: Apollo Scientific

PC210102-500MG
APOSPC210102-500MGEA 2650 NOK
APOSPC210102-500MG
(4-fluorophenyl)(pyridin-4-yl)methanol
(4-fluorophenyl)(pyridin-4-yl)methanol
Formel: C₁₂H₁₀FNO
MW: 203,212 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03840639
CAS-nummer: 65214-62-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR