1-Bromo-3-fluoropropan-2-ol 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-Bromo-3-fluoro-2-hydroxypropane

PC2445-1G
APOSPC2445-1GEA 8850 NOK
APOSPC2445-1G
1-Bromo-3-fluoropropan-2-ol 98%
1-Bromo-3-fluoropropan-2-ol
Formel: C₃H₆BrFO
MW: 156,982 g/mol
Kokepunkt: 78…80 °C (30 mm Hg)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00045119
CAS-nummer: 2107-08-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR