3,4-Difluorobenzoyl chloride 98%

Leverandør: Apollo Scientific
Danger

Synonymer: 3,4-difluorobenzoylchloride

PC2727-100G
APOSPC2727-100GEA 1775 NOK
APOSPC2727-100G
3,4-Difluorobenzoyl chloride 98%
3,4-Difluorobenzoyl chloride
Formel: C₇H₃ClF₂O
MW: 176.55 g/mol
Kokepunkt: 185…186 °C
Tetthet: 1.402 g/cm³ (25 °C)
Flash punkt: 78 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009930
CAS-nummer: 76903-88-3
EINECS: 278-570-7
FN: 3265
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR